Learn More
NPHS1 Monoclonal Antibody (OTI12D7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813415
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
NPHS1 Monoclonal antibody specifically detects NPHS1 in Human samples. It is validated for Western BlotSpecifications
| NPHS1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O60500 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4868 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI12D7 | |
| Unconjugated | |
| Nphs1 | |
| CNF; Nephrin; nephrin 1; NephrinB; nephrosis 1 homolog, nephrin; nephrosis 1, congenital, Finnish type (nephrin); nephrosis 1, nephrin; NPHN; NPHS1; NPHS1 nephrin; NPHS1, nephrin; renal glomerulus-specific cell adhesion receptor | |
| Human recombinant protein fragment corresponding to amino acids 1077-1241 of human NPHS1 (NP_004637) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.