Learn More
NQO1 Mouse anti-Human, Unconjugated, Clone: 1E5G7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67240-1-IG-20UL
Description
The NQO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NQO1. This antibody recognizes human, rat, mouse, pig antigen. The NQO1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| NQO1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry, ELISA | |
| 1E5G7 | |
| Unconjugated | |
| 20 μL | |
| 1728 | |
| IgG2a |