missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR1H3 Mouse anti-Human, Rat, Clone: 4A4F7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60134-1-IG-20UL
This item is not returnable.
View return policy
Description
The liver X receptors, LXRA and LXRB (NR1H2), form a subfamily of the nuclear receptor superfamily and are key regulators of macrophage function, controlling transcriptional programs involved in lipid homeostasis and inflammation. The inducible LXRA is highly expressed in liver, adrenal gland, intestine, adipose tissue, macrophages, lung, and kidney, whereas LXRB is ubiquitously expressed. Ligand-activated LXRs form obligate heterodimers with retinoid X receptors and regulate expression of target genes containing LXR response elements.Specifications
| NR1H3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13133, Q62685 | |
| Nr1h3 | |
| NR1H3 Fusion Protein Ag6184 | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 4A4F7 | |
| Unconjugated | |
| NR1H3 | |
| Liver X receptor alpha, LXR a, LXRA, NR1H3, Oxysterols receptor LXR alpha, RLD 1 | |
| Mouse | |
| Caprylic Acid Precipitation | |
| RUO | |
| 10062, 58852 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction