missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR1I3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21042-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the nuclear receptor superfamily, and is a key regulator of xenobiotic and endobiotic metabolism. The protein binds to DNA as a monomer or a heterodimer with the retinoid X receptor and regulates the transcription of target genes involved in drug metabolism and bilirubin clearance, such as cytochrome P450 family members. Unlike most nuclear receptors, this transcriptional regulator is constitutively active in the absence of ligand but is regulated by both agonists and inverse agonists.Specifications
| NR1I3 | |
| Polyclonal | |
| Unconjugated | |
| NR1I3 | |
| CAR, CAR BETA, CAR1, Constitutive active response, MB67, NR1I3, Orphan nuclear receptor MB67 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12355, 65035, 9970 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35627, Q14994, Q9QUS1 | |
| Nr1i3 | |
| NR1I3 Fusion Protein Ag15378 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction