Learn More
Invitrogen™ NR2F1 Recombinant Rabbit Monoclonal Antibody (24GB430)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557134
This item is not returnable.
View return policy
Description
NR2F1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene is a nuclear hormone receptor and transcriptional regulator. The encoded protein acts as a homodimer and binds to 5'-AGGTCA-3' repeats. Defects in this gene are a cause of Bosch-Boonstra optic atrophy syndrome (BBOAS).
Specifications
| NR2F1 | |
| Recombinant Monoclonal | |
| 0.65 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P10589, Q60632 | |
| Nr2f1 | |
| A synthesized peptide derived from human COUP TF1 (aa 1-53). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB430 | |
| Unconjugated | |
| Nr2f1 | |
| avian erythroblastic leukemia viral (v-erb-a) oncogene homolog-like 3; BBOAS; BBSOAS; chicken ovalbumin upstream promoter-transcription factor I; COUP transcription factor 1; COUP transcription factor I; COUP-TF I; COUP-TF1; COUPTFA; COUP-TFI; EAR3; EAR-3; Erbal3; Nr2f1; NR2F2; Nuclear receptor subfamily 2 group F member 1; nuclear receptor subfamily 2, group F, member 1; SVP44; Tcfcoup1; TFCOUP1; transcription factor COUP 1 (chicken ovalbumin upstream promoter 1, v-erb-a homolog-like 3); V-erbA-related protein 3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13865, 7025, 81808 | |
| -20°C | |
| Liquid |