missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRSF Antibody (CL0381) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42048MFV610
This item is not returnable.
View return policy
Description
NRSF Monoclonal antibody specifically detects NRSF in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| NRSF | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| Neural-restrictive silencer factor, NRSFneuron restrictive silencer factor, RE1-silencing transcription factor, X2 box repressor, XBRrepressor binding to the X2 box | |
| This antibody was developed using a recombinant protein derived from Q13127, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Stem Cell Markers, Transcription Factors and Regulators | |
| 5978 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL0381 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction