missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSUN6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17240-1-AP-20UL
This item is not returnable.
View return policy
Description
NSun6 is an RNA methyltransferase that catalyzes the methylation at position cytosine C72 in some tRNAs. NSUN6 has been reported to localize to the cytoplasm and largely colocalize with Golgi apparatus and pericentriolar matrix, indicating that NSUN6 modifies tRNAs in a late step in their biogenesis. NSun6 may also play a major role in bone metastasis and could be a valuable therapeutic target for bone metastasis.Specifications
| NSUN6 | |
| Polyclonal | |
| Unconjugated | |
| NSUN6 | |
| 4933414E04Rik, NOPD1, NSUN6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 221078, 307148, 74455 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7TS68, Q8TEA1 | |
| Nsun6 | |
| NSUN6 Fusion Protein Ag11084 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction