missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5DC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24102-1-AP-150UL
This item is not returnable.
View return policy
Description
While the exact function of the protein encoded by this gene is not known, it belongs to the 5′(3′)-deoxyribonucleotidase family.Specifications
| NT5DC1 | |
| Polyclonal | |
| Unconjugated | |
| NT5DC1 | |
| C6orf200, LP2642, NT5C2L1, NT5DC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 221294 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5TFE4 | |
| NT5DC1 | |
| NT5DC1 Fusion Protein Ag20916 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction