missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NTNG2 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148624
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
NTNG2 promotes neurite outgrowth of both axons and dendrites. Involved in controlling patterning and neuronal circuit formation at the laminar, cellular, subcellular and synaptic levels. Promotes neurite outgrowth of both axons and dendrites.
Specifications
| NTNG2 | |
| Polyclonal | |
| Unconjugated | |
| Ntng2 | |
| 2610016D08Rik; bA479K20.1; bA479K20.1 (novel protein); KIAA1857; laminet 2; laminet-2; LHLL9381; Lmnt2; LOW QUALITY PROTEIN: netrin-G2; netrin G1; netrin G2; netrin-G2; NTNG1; NTNG2; UNQ9381/PRO34206 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 171171 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8R4F1 | |
| Ntng2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Netrin-G2a, Asp18-Arg504. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction