missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUCKS1 Polyclonal Antibody

NUCKS1 Polyclonal Antibody
Brand: Invitrogen PA526535
This item is not returnable.
View return policy
Description
NUCKS1 Polyclonal Antibody for Western Blot, Flow
Specifications
| NUCKS1 | |
| Polyclonal | |
| Unconjugated | |
| Nucks1 | |
| 2700010L10Rik; 8430423A01Rik; AI647518; C78391; cyclin-dependent kinase substrate; JC7; Nucks; NUCKS1; nuclear casein kinase and cyclin dependent kinase substrate 1; nuclear casein kinase and cyclin-dependent kinase substrate 1; nuclear ubiquitous casein and cyclin-dependent kinase substrate 1; nuclear ubiquitous casein and cyclin-dependent kinases substrate; nuclear ubiquitous casein kinase 2; nuclear ubiquitous casein kinase and cyclin-dependent kinase substrate; nuclear, casein kinase and cyclin-dependent kinase substrate; P1; potential LAG1 interactor | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 64710 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.3 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9H1E3 | |
| Nucks1 | |
| KLH conjugated synthetic peptide between 45-73 amino acids from the N-terminal region of human NUCKS1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction