missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUDT19 Recombinant Rabbit Monoclonal Antibody (24GB150)

Rabbit Recombinant Monoclonal Antibody
146.00€ - 500.00€
Specifications
| Antigen | NUDT19 |
|---|---|
| Clone | 24GB150 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276324
![]() |
Invitrogen™
MA557141 |
100 μL |
500.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275807
|
Invitrogen™
MA557142 |
20 μL |
146.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NUDT19 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
NUDT19 is a coenzyme A diphosphatase that mediates the hydrolysis of a wide range of CoA esters, including choloyl-CoA and branched-chain fatty-acyl-CoA esters. At low substrate concentrations medium and long-chain fatty-acyl-CoA esters, NUDT19 are the primary substrates.Specifications
| NUDT19 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| NUDT19 | |
| CTC-379B2.4; nucleoside diphosphate-linked moiety X motif 19; nucleoside diphosphate-linked moiety X motif 19, mitochondrial; nudix (nucleoside diphosphate linked moiety X)-type motif 19; nudix hydrolase 19; nudix motif 19; NUDT19; RP2 | |
| NUDT19 | |
| Primary | |
| Affinity chromatography |
| 24GB150 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A8MXV4 | |
| 390916 | |
| A synthesized peptide derived from human NUDT19 (aa 60-190). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title