missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT2 Rabbit anti-Human, Clone: 6, Novus Biologicals™
Rabbit Monoclonal Antibody
367.00€ - 552.00€
Specifications
| Antigen | NUDT2 |
|---|---|
| Clone | 6 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18273406
|
Novus Biologicals
NBP2-90121-50UL |
50 μg |
367.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18277077
|
Novus Biologicals
NBP2-90121-100UL |
100 μg |
552.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NUDT2 Monoclonal specifically detects NUDT2 in Human samples. It is validated for ELISA.Specifications
| NUDT2 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Ap4A hydrolase, Ap4A hydrolase 1, Ap4Aase, APAH1Diadenosine 5′-5′′′-P1, bis(5′-nucleosyl)-tetraphosphatase (asymmetrical), bis(5′-nucleosyl)-tetraphosphatase [asymmetrical], diadenosine 5′-5′′-P1, Diadenosine tetraphosphatase, EC 3.6.1.17, MGC10404, Nucleoside diphosphate-linked moiety X motif 2, nudix (nucleoside diphosphate linked moiety X)-type motif 2, Nudix motif 2, P4-tetraphosphate asymmetrical hydrolase, P4-tetraphosphate pyrophosphohydrolase | |
| Recombinant Human NUDT2 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 6 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Apoptosis | |
| 0.2 um filtered solution in PBS | |
| 318 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title