Learn More
Syngene NuGenius9 XE
Product Code. 31749207 Shop All Syngene Products
Change view
Click to view available options
Pixel Size:
9 MP
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pixel Size Unit Size
31749207 9 MP 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 31749207 Supplier Syngene Supplier No. Nugenius9XE
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
NuGenius XE offers flexible DNA, stain free, autorads analysis. Intuitive touch screen system. Compact benchtop size with built in PC.
  • NuGenius XE offers flexible DNA, stain free, autorads analysis.
  • Intuitive touch screen system.
  • Compact benchtop size with built in PC.
  • Internal lighting options includes a UV transilluminator (purchased separately) for working with DNA gels (Ethidium bromide).
  • Our new UV-blue light converter screen (purchased separately) allows imaging of a wide range of safe dyes.
  • A visible converter option (purchased separately) can quickly extend its use for working with visible gels and blots (Coomassie blue and Silver stain).
  • The overhead LED white lighting is included as standard for easy sample positioning and focussing.
  • Safely excise your DNA bands from an agarose gel with the darkroom door open using the optional safety shield protecting the user when working with UV.
  • With a 5MP camera and f/1.2 motor driven lens provides exquisite pixel resolution and unrivalled sensitivity for gels and blots with a viewing area of 20 x 24 cm
  • Images are saved directly to USB or network for easy transfer.
  • Quickly generate quantitative data with GeneTools analysis software.
  • NuGenius XE comes with unlimited copies of GeneTools.
Specifications
Pixel Size 9 MP
For Use With (Application) Gel Documentation
Product Type Gel Imaging System
Display Type Touchscreen