missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NuMA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 18 publications
Brand: Novus Biologicals NB500-174
This item is not returnable.
View return policy
Description
NuMA Polyclonal specifically detects NuMA in Human, Mouse, Rat, Mammal, Marsupial, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| NuMA | |
| Polyclonal | |
| Western Blot 1:1000, Simple Western 1:500, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature | |
| Q14980 | |
| NUMA1 | |
| Full-length recombinant human NuMA expressed in insect Sf9 cells using baculovirus expression system, then purified to near homogeneity prior to immunization. | |
| 0.1 mL | |
| Breast Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Mitotic Regulators | |
| 4926 | |
| Human, Mouse, Rat, Mammalia, Marsupial, Primate | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Whole antisera with 0.02% Sodium Azide | |
| centrophilin stabilizes mitotic spindle in mitotic cells, nuclear mitotic apparatus protein 1, NUMA, NuMA protein, SP-H antigen, structural nuclear protein | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Reactive to most mammalian species tested. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion