missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Numb Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98717-100ul
This item is not returnable.
View return policy
Description
Numb Polyclonal antibody specifically detects Numb in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Numb | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 4-8 μL/mg of lysate | |
| h-Numb, numb (Drosophila) homolog, numb homolog (Drosophila), protein numb homolog, Protein S171, S171 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Numb. | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
| 8650 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction