missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Numb Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30928-25ug
This item is not returnable.
View return policy
Description
Numb Polyclonal antibody specifically detects Numb in Human samples. It is validated for Western Blot
Specifications
| Numb | |
| Polyclonal | |
| Western Blot : 1-5 μg/mL | |
| h-Numb, numb (Drosophila) homolog, numb homolog (Drosophila), protein numb homolog, Protein S171, S171 | |
| Synthetic peptide derived from human Numb protein | |
| 25 μg | |
| Cancer, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
| 8650 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction