Learn More
Thermo Scientific™ Nunc™ Cell Factory™ System Accessories
Product Code. 15778032
Click to view available options
Product Type:
Adapter Closure
Airvent Filter
Airvent Filter, 0.22 μm
Cover Cap
FEP Connector
Filter Adapter Cap, Sterile
Filter Assembly, 200 cm⊃2
Filter Assembly, 37 mm
HDPE Connector
PC Connector
System Start-Up Kit
Tubing Set, Female MPC
Tubing Set, Male MPC
Vent Cap
Vent/Close Cap
For Use With (Equipment):
Cell Factory Systems (All)
EasyFill Cell Factory Systems
Unit Size:
Each
Pack of 10
Pack of 100
Pack of 12
Pack of 2
Pack of 20
Pack of 40
Product Code. 15778032
Brand: Thermo Scientific™ 140817
Only null left
Add to basket
Edge
Add to basket
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories.
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
Connections made easy
  • Plug a media bag or tubing set into any cell factory
Save time and reduce costs
  • Cleaning processes and sterility validations are minimized
  • Most accessories are gamma irradiated and individually wrapped (Note: Connector Cat. Nos- 140099 and 173250 are not available gamma irradiated)
Prevent buildup of back pressure during filling application
  • Venting options provide additional air venting when filling and harvesting by gravity or with a peristaltic pump
TRUSTED_SUSTAINABILITY
Specifications
Description Tubing Set with Male MPC connector
For Use With (Equipment) Cell Factory Systems (All)
No. per Pack 1
Product Type Tubing Set, Male MPC
Sterility Sterile
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.