Learn More
Thermo Scientific™ Nunc™ IVF ICSI Dish
Beskrivning
Experience consistently reliable, high-quality results during fertilization. Thermo Scientific™ Nunc IVF ICSI Dishes meet the exacting requirements of IVF testing, certification, and validation.
Innovative features for quality and ease-of-use
- Maintain optimal temperature with true flat bottom
- Low profile provides optimal angles during ICSI process
- Clearly see sample with super polished optical center
- Increased safety with easy grip design
- Certificates of conformity available for every lot
Rigorously tested, certified and validated for IVF
- CE-marked (Notified Body 2797), FDA-listed
- Produced under ISO 13485 with full lot traceability
- Certificates of conformity for your assurance on every lot
Sterile (SAL 10-6)
- Non-pyrogenic (LAL test)
- USP Class VI test
- Lot-release tested by 1-cell stage Mouse Embryo Assay (MEA) and Human Sperm Survival Assay (HSSA)
Specifikationer
Specifikationer
| Certifications/Compliance | CE-marked, FDA-cleared, MEA-tested |
| Description | IVF ICSI Dish, Lid w/ Airvent |
| Bottom Shape | Flat |
| Color | Clear |
| Material | Polystyrene (USP Class VI) |
| Sterility | Sterile |
| Surface Treatment | Non-treated |
| Type | Dish |
| Well Count | 1 |
| No. per Case | 120 |
| Visa mer |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.