missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ NUP50 Overexpression Lysate

Product Code. 18682889 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18682889 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18682889 Supplier Novus Biologicals™ Supplier No. NBP307978
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Overexpression Lysate

TRUSTED_SUSTAINABILITY

Specifications

Preparation Method This product is intended for use as a positive control in Western Blot. Each vial of cell lysate contains 100 ug of total protein which should be sufficient for 20-50 reactions. Depending on over-expression level, antibody affinity and detection system, some lysates can go as low as 0.1 μL / load. We recommend starting with 5 ug of cell lysate. Add an equal amount of cell lysate and 2 x SDS Sample buffer and boil the SDS samples for 10 minutes before loading.
Quantity 0.1 mg
Host Species Human
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Target NUP50
Synonym 50 kDa nucleoporin, MGC39961, NPAP60, NPAP60LNucleoporin Nup50, nuclear pore complex protein Nup50, Nuclear pore-associated protein 60 kDa-like, nuclear pore-associated protein 60 L, nucleoporin 50 kD, nucleoporin 50 kDa
Gene Symbol NUP50
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.