Learn More
Thermo Scientific™ Nutrient Agar (Dehydrated)
Product Code. 12754126
Change view
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12754126 5 kg 5kg
10219252 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12754126 Supplier Thermo Scientific™ Supplier No. CM0003T
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Cultivate and isolate fastidious pathogens and other micro-organisms with this nutritive, non-selective medium.
Cultivate and isolate fastidious pathogens and other microorganisms with Thermo Scientific™ Oxoid™ Nutrient Agar (Dehydrated). This non-selective medium contains beef extracts combined with peptone, yeast extracts and sodium chloride to provide the nitrogen compounds, carbon, vitamins and some trace ingredients necessary for the growth of bacteria.
Nutrient Agar is a basic culture medium used to subculture organisms for maintenance purposes or to check the purity of subcultures from isolation plates prior to biochemical or serological tests.
In semi-solid form, as agar slopes or agar butts, the medium is used to maintain control organisms1.
Nutrient Agar is suitable for teaching and demonstration purposes. It contains a concentration of 1.5% agar to permit the addition of up to 10% blood or other biological fluid, as required. The medium, without additions, may be used for the cultivation of organisms which are not exacting in their nutritional requirements.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: AFNOR, AOAC, BSI, FDA, ISO, NMKL.
Form Powder
Product Type Nutrient Agar
Quantity 5 kg
For Laboratory Use Only