Description
- Use these fittings for a variety of critical applications, including syringes or small tubing
- All parts meet tight dimensional tolerances for increased reliability and process purity
- Choose from a range of sizes, with a variety of connection options
- Good chemical resistance. Max temperature 235°C (455°F)
- Sterilize by autoclave, ethylene oxide, or gamma irradiation
- All female ports may be used with either slip or luer lock connections
- Plugs and caps are also available
Specifications
Specifications
| Autoclavable | Autoclavable |
| Packaging Quantity | 25/Pk. |
| Max. Operating Temperature (Metric) | 235°C |