Learn More
Brady™ Nylon Thermal Transfer Printer Labels
Product Code. 10273143 Shop All Brady Products
Change view
Click to view available options
Color:
White
Yellow
Size (Metric):
12.7 mm dia.
12.7 x 25.4 mm
12.7W x 25.4 mmH
22.86W x 38.1 mmH
25.4W x 36.5 mmH
25.4W x 66.68 mmH
41.28W x 15.24 mmH
9.53 mm dia.
Quantity:
1000/Pk.
1500/Pk.
2500/Pk.
3000/Pk
3000/Pk.
Unit Size:
1 roll
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Size (Metric) Quantity Unit Size
10273143 White 12.7 x 25.4 mm 3000/Pk. 1 roll
11806023 White 41.28W x 15.24 mmH 3000/Pk. 1 roll
11884664 Yellow 12.7 mm dia. 1000/Pk. 1 roll
10395443 White 25.4W x 66.68 mmH 2500/Pk. 1 roll
11886023 White 25.4W x 36.5 mmH 1500/Pk. 1 roll
11819734 Yellow 9.53 mm dia. 3000/Pk. 1 roll
10671344 White 9.53 mm dia. 3000/Pk 1 roll
10415992 White 12.7W x 25.4 mmH 1000/Pk. 1 roll
10051313 White 12.7 mm dia. 3000/Pk. 1 roll
10192752 White 22.86W x 38.1 mmH 3000/Pk. 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10273143 Supplier Brady™ Supplier No. 217075
  • ONLINE EXCLUSIVE PRICE
    395.00€
    347.48€ / 1 rolls
    Save 47.52€
    12% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For smooth, textured/rough or Highly textured/Low surface energy surfaces.
  • Permanent acrylic adhesive
  • Matt finish
  • Nylon cloth labels
Specifications
Autoclavable Autoclavable
Certifications/Compliance UL Recognized, CSA Accepted, Halogen Free (DIN VDE 0472/part 815) Approvals are for White material only, Compliant with Directive 2011/65/EU and 2015/863
Description Thermal Transfer Printable Labels
Adhesive Type Acrylic, Permanent
Coating None
Color White
Display Blank
Finish Matte
Format Die-cut
Material Nylon
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Thermal Transfer Printer Label
Sterility Non-sterile
For Use With (Application) Component and equipment labelling, Electrical labelling, General identification, Laboratory labelling, Warehouse marking, Wire and cable labelling
For Use With (Equipment) BBP11, BBP12, PR Plus, THT 76 mm Core Printers
Labels per Row 3
Min. Operating Temperature (Metric) -40°C
No. per Pack 3000 Labels/Roll
Quantity 3000/Pk.
Shape Rectangle
Size (Metric) 12.7 x 25.4 mm
Temperature Range (English) -40°F to +194°F
Temperature Range (Metric) -40°C to +90°C
Thickness (English) 0.0065 in.
Thickness (Metric) 0.166 mm
Vertical Repeat (Metric) 28.40 mm
Dimensions (L x W) 25.4 x 12.70 mm
Length (English) 1 in.
Width (English) 0.5 in.
Width (Metric) 12.70 mm
Height (Metric) 25.40 mm
Alternate Part Number BPT-611-499