Learn More
Masterflex™ Nylon Threaded Fittings
Product Code. 11745768
Change view
Click to view available options
For Use With:
0.125 in. NPT(M)
0.125in.
0.25 in. NPT(M)
0.25in. Tubing
0.375 in. NPT(M)
0.375in.
0.75 x 0.5in.
0.75in.
Unit Size:
Pack of 10
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
11745768 0.125 in. NPT(M) Pack of 10
11770369 0.25 in. NPT(M) Pack of 10
11751084 0.25in. Tubing Pack of 6
11780369 0.375 in. NPT(M) Pack of 10
11711104 0.375in. Pack of 6
11791084 0.125in. Pack of 6
11731104 0.75in. Pack of 6
11701154 0.75 x 0.5in. Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11745768 Supplier Masterflex™ Supplier No. 3170191
  • ONLINE EXCLUSIVE PRICE
    10.05€
    8.87€ / Pack of 10
    Save 1.18€
    11% Off
In Stock
Add to basket
Edge
Add to basket
Specifications
Material Nylon
For Use With 0.125 in. NPT(M)
Min. Operating Temperature (Metric) -40°C
Packaging Quantity 10 Pack
Max. Operating Temperature (Metric) 93°C
Product Type Threaded Fitting