missing translation for 'onlineSavingsMsg'
Learn More

O(1),O(3)-Bis(ethoxycarbonylmethyl)-p-tert-butylcalix[4]arene, Thermo Scientific™

Product Code. 11464147
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464147 0.5 g 0.5g
11454147 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464147 Supplier Thermo Scientific Alfa Aesar Supplier No. A19594.02
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
O(1),O(3)-Bis(ethoxycarbonylmethyl)-p-tert-butylcalix[4]arene is used as an intermediates, amino acid and other fine chemicals.

Solubility
Sparingly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97600-49-2
Molecular Formula C52H68O8
Molecular Weight (g/mol) 821.108
MDL Number MFCD00798575
InChI Key RCVLXPWJVXHCMD-UHFFFAOYSA-N
Synonym o 1 ,o 3-bis ethoxycarbonylmethyl-p-tert-butylcalix 4 arene, o 1 ,o 3-bis carbethoxymethyl-p-tert-butylcalix 4 arene, 2,6-5-tert-butyl-2-hydroxy-1,3-phenylene bis methylene 5-tert-butyl-2-ethoxycarbonylmethoxy-1,3-phenylene methylene-4-tert-butylphenol, ethyl 2-5,11,17,23-tetra-tert-butyl-27-2-ethoxy-2-oxoethoxy-26,28-dihydroxypentacyclo 19.3.1.1?,?.1?,??.1??,?? octacosa-1 25 ,3 28 ,4,6,9,11,13 27 ,15 26 ,16,18,21,23-dodecaen-25-yl oxy acetate, ethyl 2-5,11,17,23-tetra-tert-butyl-27-2-ethoxy-2-oxoethoxy-26,28-dihydroxypentacyclo 19.3.1.1_,?.1?,__.1_?,_? octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15 26 ,16,18,21,23-dodecaen-25-yl oxy acetate
PubChem CID 10887342
SMILES CCOC(=O)COC1=C2CC3=CC(=CC(=C3O)CC4=C(C(=CC(=C4)C(C)(C)C)CC5=C(C(=CC(=C5)C(C)(C)C)CC1=CC(=C2)C(C)(C)C)O)OCC(=O)OCC)C(C)(C)C

Specifications

Melting Point 182°C to 184°C
Quantity 0.5 g
Solubility Information Sparingly soluble in water.
Formula Weight 821.11
Chemical Name or Material O(1),O(3)-Bis(ethoxycarbonylmethyl)-p-tert-butylcalix[4]arene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.