missing translation for 'onlineSavingsMsg'
Learn More

O-[4-(Aminomethyl)benzyl]guanine, TRC

Product Code. 30137208
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137208 100 mg 100mg
30136882 2.5 g 2.5g
30008792 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137208 Supplier Toronto Research Chemicals Supplier No. TRCA614425100MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 674799-96-3
Molecular Formula C13 H14 N6 O
Molecular Weight (g/mol) 270.29
Synonym 9H-Purin-2-amine, 6-[[4-(aminomethyl)phenyl]methoxy]-, 1H-Purin-2-amine, 6-[[4-(aminomethyl)phenyl]methoxy]- (9CI), 6-[[4-(Aminomethyl)phenyl]methoxy]-9H-purin-2-amine, O-[4-(Aminomethyl)benzyl]guanine
IUPAC Name 6-[[4-(aminomethyl)phenyl]methoxy]-9H-purin-2-amine
SMILES NCc1ccc(COc2nc(N)nc3[nH]cnc23)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H14N6O/c14-5-8-1-3-9(4-2-8)6-20-12-10-11(17-7-16-10)18-13(15)19-12/h1-4,7H,5-6,14H2,(H3,15,16,17,18,19)
Formula Weight 270.1229
Percent Purity >95%
Chemical Name or Material O6-[4-(Aminomethyl)benzyl]guanine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.