missing translation for 'onlineSavingsMsg'
Learn More

O-Acetylsalicyloyl Chloride, TRC

Product Code. 30314315
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314315 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314315 Supplier Toronto Research Chemicals Supplier No. TRCA18793510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O-Acetylsalicyloyl Chloride
CAS 5538-51-2
Molecular Formula C9H7ClO3
Quantity 10 g
Synonym 2-(Acetyloxy)benzoyl Chloride, Salicyloyl Chloride Acetate, 2-(Acetyloxy)benzoyl Chloride, 2-(Chlorocarbonyl)phenyl Acetate, 2-Acetoxybenzoyl Chloride, 2-Acetylsalicyloyl Chloride, Acetic Acid 2-Chlorocarbonylphenyl Ester, Acetoxybenzoic Acid Chloride, Acetylsalicyl Chloride, Acetylsalicylic Acid Chloride, Acetylsalicyloyl Chloride, Aspiryl Chloride, NSC 97216, O-Acetylsalicylic Acid Chloride, o-(Acetyloxy)benzoyl Chloride, o-Acetoxybenzoyl Chloride
SMILES CC(=O)Oc1ccccc1C(=O)Cl
Molecular Weight (g/mol) 198.6
Formula Weight 198.0084
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.