missing translation for 'onlineSavingsMsg'
Learn More

O-Benzylhydroxylamine, 96%

Product Code. 11383167
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383167 5 g 5g
11393167 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383167 Supplier Thermo Scientific Alfa Aesar Supplier No. L13601.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 622-33-3 | C7H9NO | 123.155 g/mol

O-benzylhydroxylamine used in the synthesis of alfa- hydroxybenzylamines from alfa-hydroxyketones. It is also involved in the preparation of hydroxylamines and hydroxyamates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-benzylhydroxylamine used in the synthesis of alfa- hydroxybenzylamines from alfa-hydroxyketones. It is also involved in the preparation of hydroxylamines and hydroxyamates.

Solubility
Soluble in dimethylsulfoxide and methanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-33-3
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.61
MDL Number MFCD00221709
InChI Key HYDZPXNVHXJHBG-UHFFFAOYSA-N
Synonym benzyloxyamine, hydroxylamine, o-phenylmethyl, hydroxylamine, o-benzyl, o-benzyl-hydroxylamine, benzyl-o-hydroxylamine, oxybenzylamine, o-phenylmethyl hydroxylamine, o-benzyloxylamine, phenylmethoxyamine, benzyloxy amine
PubChem CID 102313
IUPAC Name hydrogen O-benzylhydroxylamine chloride
SMILES [H+].[Cl-].NOCC1=CC=CC=C1

Specifications

Density 1.058
Boiling Point 118°C to 119°C (30 mmHg)
Linear Formula C6H5CH2ONH2
Refractive Index 1.541
Quantity 5 g
UN Number UN2735
Beilstein 1906691
Solubility Information Soluble in dimethylsulfoxide and methanol.
Formula Weight 123.15
Percent Purity 96%
Chemical Name or Material O-Benzylhydroxylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.