missing translation for 'onlineSavingsMsg'
Learn More

O-Isopropyl Ethylthiocarbamate, TRC

Product Code. 30422129
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422129 10 g -
30414901 5 g 5g
30416253 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30422129 Supplier Toronto Research Chemicals Supplier No. TRCO24744010G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-98-0
Molecular Formula C6 H13 N O S
Molecular Weight (g/mol) 147.238
Synonym Carbamic acid, ethylthio-, O-isopropyl ester (6CI,8CI), Carbamothioic acid, ethyl-, O-(1-methylethyl) ester (9CI), O-(1-Methylethyl) N-ethylcarbamothioate, Ethylthiocarbamic acid, O-isopropyl ester, Minerec 2030, O-Isopropyl N-ethylthiocarbamate, O-Isopropyl N-ethylthionocarbamate, O-Isopropyl ethylthiocarbamate, XKP 03, Z 200, Z 200 (flotation agent)
IUPAC Name O-propan-2-yl N-ethylcarbamothioate
SMILES CCNC(=S)OC(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H13NOS/c1-4-7-6(9)8-5(2)3/h5H,4H2,1-3H3,(H,7,9)
Formula Weight 147.0718
Chemical Name or Material N-Ethyl-O-isopropylthiocarbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.