missing translation for 'onlineSavingsMsg'
Learn More

O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98+%, Thermo Scientific Chemicals

Product Code. 15450676
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450676 5 g 5g
15440676 1 g 1g
15460676 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15450676 Supplier Thermo Scientific Alfa Aesar Supplier No. H54035.06
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
This item is not returnable. View return policy

Coupling reagent for peptide synthesis and the formation of other amides. Reactant for synthesis of liposomal contrast agents for magnetic resonance imaging, synthesis of protein labeling molecules, synthesis of thiol-reactive Cy5 derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Coupling reagent for peptide synthesis and the formation of other amides. Reactant for synthesis of liposomal contrast agents for magnetic resonance imaging, synthesis of protein labeling molecules, synthesis of thiol-reactive Cy5 derivatives.

Solubility
Soluble in acetonitrile 0.5 g/mL.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 265651-18-1
Molecular Formula C9H16F6N3O3P
Molecular Weight (g/mol) 359.21
MDL Number MFCD01863753
InChI Key STWZCCVNXFLDDD-UHFFFAOYSA-N
Synonym hstu, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate, hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate, ambotzrl-1039, o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate
PubChem CID 16211151
IUPAC Name [dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C

Specifications

Melting Point ∽220°C (decomposition)
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in acetonitrile 0.5g/mL.
Formula Weight 359.21
Percent Purity ≥98%
Chemical Name or Material O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.