Learn More
o-Nitrofluorobenzene, TRC
Product Code. 30132696
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132696 50 g 50g
30138473 100 g 100g
30003672 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132696 Supplier Toronto Research Chemicals Supplier No. TRCN49592050G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1493-27-2
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.1
Synonym 1-Fluoro-2-nitrobenzene, 2-Fluoro-1-nitrobenzene, 2-Fluoronitrobenzene, 2-Nitro-1-fluorobenzene, 2-Nitrofluorobenzene, 4-Fluoro-3-nitrobenzene, NSC 51869, o-Fluoronitrobenzene;
IUPAC Name 1-fluoro-2-nitrobenzene
SMILES [O-][N+](=O)c1ccccc1F
Specifications
Quantity 50 g
InChI Formula InChI=1S/C6H4FNO2/c7-5-3-1-2-4-6(5)8(9)10/h1-4H
Formula Weight 141.0226
Chemical Name or Material o-Nitrofluorobenzene