missing translation for 'onlineSavingsMsg'
Learn More

O,O,S-Triethyl Phosphorothiolate, TRC

Product Code. 30012572
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012572 2.5 g 2.5g
30010246 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012572 Supplier Toronto Research Chemicals Supplier No. TRCT7770152.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1186-09-0
Molecular Formula C6 H15 O3 P S
Molecular Weight (g/mol) 198.22
Synonym Phosphorothioic acid, O,O,S-triethyl ester, Ethyl phosphorothioate ((EtO)2(EtS)PO) (6CI), Ethyl thiophosphate ((EtO)2(EtS)PO) (5CI), O,O,S-Triethyl phosphorothioate, O,O,S-Triethyl phosphorothiolate, O,O,S-Triethyl thiophosphate, O,O-Diethyl S-ethyl phosphorothioate, Thiophosphoric acid O,O,S-triethyl ester, Triethyl phosphorothioate
IUPAC Name 1-[ethoxy(ethylsulfanyl)phosphoryl]oxyethane
SMILES CCOP(=O)(OCC)SCC

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C6H15O3PS/c1-4-8-10(7,9-5-2)11-6-3/h4-6H2,1-3H3
Formula Weight 198.048
Chemical Name or Material O,O,S-Triethyl Phosphorothiolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.