missing translation for 'onlineSavingsMsg'
Learn More

o-Phthalaldehyde, Serva Electrophoresis™

Product Code. 11781558
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11781558 5 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11781558 Supplier Serva Electrophoresis Supplier No. 3280001
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 643-79-8
Melting Point 54°C to 57°C
Melting Range 54°C to 57°C
Molecular Formula C8H6O2
MDL Number MFCD00003335
Quantity 5 g
Synonym o-phthalaldehyde, o-phthaldialdehyde, 1,2-benzenedicarboxaldehyde, benzene-1,2-dicarboxaldehyde, phthaldialdehyde, phthalic aldehyde, phthalic dialdehyde, phthalyldicarboxaldehyde, ortho-phthalaldehyde, o-phthaldehyde
InChI Key ZWLUXSQADUDCSB-UHFFFAOYSA-N
SMILES O=CC1=CC=CC=C1C=O
IUPAC Name benzene-1,2-dicarbaldehyde
Molecular Weight (g/mol) 134.13
PubChem CID 4807
ChEBI CHEBI:70851
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.