missing translation for 'onlineSavingsMsg'
Learn More

O-Tolidine Solution, Ellms Hauser, Chem Lab™

Product Code. 11959192
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11959192 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11959192 Supplier Chem-Lab Supplier No. CL02.0501.1000
  • ONLINE EXCLUSIVE PRICE
    99.00€
    50.00€ / 1 L
    Save 49.00€
    49% Off
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 119-93-7
Molecular Formula C14H16N2
Quantity 1 L
Synonym o-tolidine, 3,3'-dimethylbenzidine, orthotolidine, diaminoditolyl, 2-tolidine, diaminotolyl, bianisidine, 3,3'-dimethylbiphenyl-4,4'-diamine, 4,4'-bi-o-toluidine, 3,3'-tolidine
InChI Key NUIURNJTPRWVAP-UHFFFAOYSA-N
Molecular Weight (g/mol) 212.30
PubChem CID 8413
ChEBI CHEBI:34320
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ellm's Hauser reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H350-May cause cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: o-Tolidine, Hydrochloric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.