Learn More
Motic™ Objective, Achromatic 10X/0.25
Product Code. 15495082
Change view
Click to view available options
Magnification Power:
10X
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Magnification Power unitSize
15495082 10X Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15495082 Supplier Motic™ Supplier No. 1101001701133
  • ONLINE EXCLUSIVE PRICE
    29.85€
    26.84€ / Pack of 1
    Save 3.01€
    10% Off
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
Designed with chromatic aberration correction for 2 colors. Motic™ Objective, Achromatic 10X/0.25 is spherically corrected for one color.
  • Achromatic.
  • Chromatic aberration correction for 2 colors.
  • Spherical correction for 1 color.
Specifications
Type Achromat Objective
Description NA: 0.25
For Use With (Equipment) SFC-100, DM-111, F-1110 Microscopes
Magnification Power 10X