Learn More
Carl Zeiss™ Objective EC Plan Neofluar
Use these universal objectives for all contrast methods: bright field, dark field, phase contrast, DIC, fluorescence and Pol. Zeiss™ Objective EC Plan Neofluar feature high resolving power and brilliant, rich-contrast, completely flat images for observation and documentation.
Brand:  Carl Zeiss™ 420350-9900-000
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11875560
  • 2358.50€ / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
  • Enhanced Contrast Plan-Neofluar
  • Minimized stray light: with a visible increase in contrast important
  • High transmission of wavelengths from the near UV range (>40% starting at 340nm)
  • Glass types of low autofluorescence makes these objectives
  • Ideal for epi-fluorescence microscopy
Specifications
Specifications
Objective EC
20X
1 Ea.