Learn More
Occludin Antibody (CL1567) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52972AF700
Product Code. 30293165
  • 759.00€ / 0.10mL
Description
Occludin Monoclonal antibody specifically detects Occludin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Occludin
Monoclonal
Alexa Fluor 700
BLCPMG, occludin
This antibody was developed using a recombinant protein derived from Q16625, with the exact immunogen sequence remaining proprietary.
0.1 mL
Signal Transduction
100506658
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL1567