missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Occludin Antibody (OCLN/2183), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08880AF350
This item is not returnable.
View return policy
Description
Occludin Monoclonal antibody specifically detects Occludin in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications
| Occludin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| BLCPMG, occludin | |
| Recombinant human Occludin fragment around aa 282-415 (Exact sequence is proprietary) (Uniprot: Q16625) | |
| 100 μg | |
| Signal Transduction | |
| 100506658 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| OCLN/2183 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction