missing translation for 'onlineSavingsMsg'
Learn More

OCIAD1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

178.00€

Specifications

Antigen OCIAD1
Concentration 1 mg/mL
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18539873
View Documents
Novus Biologicals
NBP1-76242-0.025mg
0.025 mg
178.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

OCIAD1 Polyclonal specifically detects OCIAD1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

OCIAD1
Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL
Polyclonal
Rabbit
Human
NP_060300
54940
Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human OCIAD1. The immunogen is located within the last 50 amino acids of OCIAD1. Amino Acid Squence: PKKEVKVNKYGDTWDE
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
1 mg/mL
Western Blot
Unconjugated
RUO
PBS with 0.02% Sodium Azide
Asrij, MGC111072, OCIA domain containing 1, OCIA domain-containing protein 1, OCIAFLJ20455, Ovarian carcinoma immunoreactive antigen, TPA018
OCIAD1
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.