Learn More
Invitrogen™ Ocrelizumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559573
 View more versions of this product
Product Code. 30284689
  • 477.00€ / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Ocrelizumab is a CD20-directed cytolytic antibody indicated for the treatment of patients with relapsing or primary progressive forms of multiple sclerosis. It is a second-generation recombinant humanized monoclonal IgG1 antibody that selectively targets the B lymphocytes that express the CD20 antigen. As a humanized molecule, ocrelizumab is expected to be less immunogenic with repeated infusions which improves the benefit-to-risk profile for patients with relapsing or progressive forms of MS.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ocrelizumab Humanized
Recombinant Monoclonal
Unconjugated
Ocrevus
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized