missing translation for 'onlineSavingsMsg'
Learn More

Octaacetyl Xylotriose, TRC

Product Code. 30425599
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425599 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425599 Supplier Toronto Research Chemicals Supplier No. TRCO185005500MG
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octaacetyl Xylotriose
CAS 67226-05-5
Molecular Formula C31H42O21
Quantity 500 mg
Synonym O-2,3,4-Tri-O-acetyl-beta-D-xylopyranosyl-(1-4)-O-2,3-di-O-acetyl-beta-D-xylopyranosyl-(1-4)-D-xylopyranose 1,2,3-Triacetate, (3R,4S,5R)-5-(((2S,3R,4S,5R)-3,4-Diacetoxy-5-(((2S,3R,4S,5R)-3,4,5-triacetoxytetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2,3,4-triyl Triacetate
InChI Formula InChI=1S/C31H42O21/c1-12(32)43-20-9-41-30(27(48-17(6)37)23(20)44-13(2)33)52-22-11-42-31(28(49-18(7)38)25(22)46-15(4)35)51-21-10-40-29(50-19(8)39)26(47-16(5)36)24(21)45-14(3)34/h20-31H,9-11H2,1-8H3/t20-,21-,22-,23+,24+,25+,26-,27-,28-,29?,30+,31+/m1/s1
SMILES CC(=O)O[C@@H]1CO[C@@H](O[C@@H]2CO[C@@H](O[C@@H]3COC(OC(=O)C)[C@H](OC(=O)C)[C@H]3OC(=O)C)[C@H](OC(=O)C)[C@H]2OC(=O)C)[C@H](OC(=O)C)[C@H]1OC(=O)C
IUPAC Name [(3R,4S,5R,6S)-4,5-diacetyloxy-6-[(3R,4S,5R,6S)-4,5-diacetyloxy-6-[(3R,4S,5R)-4,5,6-triacetyloxyoxan-3-yl]oxyoxan-3-yl]oxyoxan-3-yl] acetate
Molecular Weight (g/mol) 750.65
Formula Weight 750.2219
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.