missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecanoic-9,9,10,10-d4 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Octadecanoic-9,9,10,10-D4 Acid |
| CAS | 130048-57-6 |
| Molecular Formula | C18 D4 H32 O2 |
| Quantity | 0.25 g |
| Synonym | Octadecanoic-9,9,10,10-d4 acid (9 CI, ACI), Stearic-9,9,10,10-d4 acid (5 CI), 9,9,10,10-Tetradeuterostearic acid, [9,9,10,10-d4]-Stearic acid, Stearic acid-9,9,10,10-d4, Octadecanoic-9,9,10,10-D4 acid |
| InChI Formula | InChI=1 S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i9D2,10D2 |
| SMILES | [2 H]C([2 H])(CCCCCCCC)C([2 H])([2 H])CCCCCCCC(=O)O |
| IUPAC Name | 9,9,10,10-tetradeuteriooctadecanoic acid |
| Molecular Weight (g/mol) | 288.502 |
| Formula Weight | 288.297 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction