missing translation for 'onlineSavingsMsg'
Learn More

OctaInd, TRC

Product Code. 30424325
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30424325 1 g
30419643 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30424325 Supplier Toronto Research Chemicals Supplier No. TRCO2371001G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1084889-51-9
Molecular Formula C18 H12 Br8
Molecular Weight (g/mol) 867.5199
Synonym OctaInd, 1H-Indene, 4,5,6,7-tetrabromo-2,3-dihydro-1,1,3-trimethyl-3-(2,3,4,5-tetrabromophenyl)-
IUPAC Name 4,5,6,7-tetrabromo-1,1,3-trimethyl-3-(2,3,4,5-tetrabromophenyl)-2H-indene
SMILES CC1(C)CC(C)(c2cc(Br)c(Br)c(Br)c2Br)c3c(Br)c(Br)c(Br)c(Br)c13

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H12Br8/c1-17(2)5-18(3,6-4-7(19)11(21)14(24)10(6)20)9-8(17)12(22)15(25)16(26)13(9)23/h4H,5H2,1-3H3
Formula Weight 859.4406
Chemical Name or Material 4,5,6,7-Tetrabromo-2,3-dihydro-1,1,3-trimethyl-3-(2,3,4,5-tetrabromophenyl)-1H-indene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.