missing translation for 'onlineSavingsMsg'
Learn More

Octanoic acid, 98+%

Product Code. 11469063
Change view
Click to view available options
Quantity:
10,000 mL
25 mL
2500 mL
500 mL
Unit Size:
10000mL
2500mL
25mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469063 2500 mL 2500mL
11428710 25 mL 25mL
11459063 500 mL 500mL
11479063 10,000 mL 10000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11469063 Supplier Thermo Scientific Alfa Aesar Supplier No. A11149.0F
  • ONLINE EXCLUSIVE PRICE
    93.90€
    84.65€ / 2500mL
    Save 9.25€
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 124-07-2 | C8H16O2 | 144.214 g/mol

Octanoic acid is used in the production of esters, which finds application in perfumery and also dyes manufacture. It is used as a food contact surface sanitizer in commercial food handling, as a dietary supplement and as disinfectant in health care facilities, recreational facilities, livestock premises and industrial facilities. Its acid chloride is used in the synthesis of perfluorooctanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octanoic acid is used in the production of esters, which finds application in perfumery and also dyes manufacture. It is used as a food contact surface sanitizer in commercial food handling, as a dietary supplement and as disinfectant in health care facilities, recreational facilities, livestock premises and industrial facilities. Its acid chloride is used in the synthesis of perfluorooctanoic acid.

Solubility
Misicible with alcohol, chloroform, ether, carbon disulfide, petroleum ether and acetonitrile.Slightly miscible with water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-07-2
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00004429
InChI Key WWZKQHOCKIZLMA-UHFFFAOYSA-N
Synonym caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid
PubChem CID 379
ChEBI CHEBI:28837
IUPAC Name octanoic acid
SMILES CCCCCCCC(=O)O

Specifications

Melting Point 16°C to 17°C
Density 0.91
pH 4
Boiling Point 236°C to 238°C
Flash Point 132°C (269°F)
Assay Percent Range >98%
Odor Rancid
Linear Formula CH3(CH2)6CO2H
Refractive Index 1.4275
Quantity 2500 mL
UN Number UN3265
Beilstein 1747180
Merck Index 14,1765
Solubility Information Misicible with alcohol,chloroform,ether,carbon disulfide,petroleum ether and acetonitrile.Slightly miscible with water.
Formula Weight 144.21
Percent Purity ≥98%
Chemical Name or Material Octanoic acid
Show More Show Less
compliance-icons
Product Identifier
  • Octanoic acid
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Octanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.