missing translation for 'onlineSavingsMsg'
Learn More

Octanoic Acid, TRC

Product Code. 30412166
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412166 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412166 Supplier Toronto Research Chemicals Supplier No. TRCO23788050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Caprylic Acid
CAS 124-07-2
Molecular Formula C8 H16 O2
Quantity 50 g
Synonym 1-Heptanecarboxylic acid, Caprylic acid, Cg 100, Cg 100 (fatty acid), Edenor 8-99MY, Edenor C 8-98-100, Emery 657, Kortacid 0899, Lunac 8-95, Lunac 8-98, NAA 82, NSC 5024, Neo-Fat 8, Neo-Fat 8S, Octylic acid, Prifac 2901, n-Caprylic acid, n-Octanoic acid, n-Octoic acid, n-Octylic acid
InChI Formula InChI=1S/C8H16O2/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3,(H,9,10)
SMILES CCCCCCCC(=O)O
IUPAC Name octanoic acid
Molecular Weight (g/mol) 144.21
Formula Weight 144.115
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.