missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octanoyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Octanoyl Chloride |
| CAS | 111-64-8 |
| Molecular Formula | C8H15ClO |
| Quantity | 10 g |
| Synonym | 1-Octanoic Acid Chloride, Caprylic Acid Chloride, Capryloyl Chloride, Caprylyl Chloride, Octanoic Acid Chloride, Octanoic Chloride, n-Octanoyl Chloride |
| InChI Formula | InChI=1S/C8H15ClO/c1-2-3-4-5-6-7-8(9)10/h2-7H2,1H3 |
| SMILES | CCCCCCCC(=O)Cl |
| IUPAC Name | octanoyl chloride |
| Molecular Weight (g/mol) | 162.66 |
| Formula Weight | 162.0811 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Haben Sie Verbesserungsvorschläge?