missing translation for 'onlineSavingsMsg'
Learn More

Octinoxate, LoGiCal

Product Code. 30464208
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464208 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464208 Supplier LoGiCal Supplier No. LGCFOR0729.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octinoxate
CAS 5466-77-3
Analyte or Component Names Octinoxate
Shipping Condition Room Temperature
Molecular Formula C18 H26 O3
Certifications/Compliance ISO 9001
Synonym Octinoxate, 2-Ethylhexyl 3-(4-methoxyphenyl)prop-2-enoate, 2-Ethylhexyl 4-methoxycinnamate, 2-Ethylhexyl methoxycinnamate, 2-Ethylhexyl p-methoxycinnamate, 3-(4-Methoxyphenyl)-2-propenoic acid 2-ethylhexyl ester, 4-Methoxycinnamic acid 2-ethylhexyl ester, Escalol 557, Escalol 557NB, Escalol 557T, Ethylhexyl methoxycinnamate, Ethylhexyl p-methoxycinnamate, Eusolex 2292, Eusolex UV-Pearls OMC, Jeescreen OMC, NSC 26466, Neo Heliopan AV, Nomcort TAB, Nomcort TAB-R, Octyl 4-methoxycinnamate, Octyl methoxycinnamate, Octyl p-methoxycinnamate, Parsol MCX, Parsol MCX-SA, Parsol MOX, Solarom OMC, Sun Caps 664, Sunscreen AV, Tinosorb OMC, Uvinul 3088, Uvinul MC 80, Uvinul MC 80N, Uvinul MC 90, p-Methoxycinnamic acid 2-ethylhexyl ester
InChI Formula InChI=1S/C18H26O3/c1-4-6-7-15(5-2)14-21-18(19)13-10-16-8-11-17(20-3)12-9-16/h8-13,15H,4-7,14H2,1-3H3/b13-10+
Solution Type Neat
SMILES CCCCC(CC)COC(=O)\C=C\c1ccc(OC)cc1
IUPAC Name 2-ethylhexyl (E)-3-(4-methoxyphenyl)prop-2-enoate
Industry Type Clinical Testing
Molecular Weight (g/mol) 290.4
Quantity 10 mg
Formula Weight 290.1882 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.