missing translation for 'onlineSavingsMsg'
Learn More

Octyl 3-Octyloxiraneoctanoic Acid, TRC

Product Code. 30135119
Change view
Click to view available options
Quantity:
100 mg
5 g
Unit Size:
100mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135119 5 g 5g
30010502 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135119 Supplier Toronto Research Chemicals Supplier No. TRCO2925005G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-84-3
Molecular Formula C26 H50 O3
Molecular Weight (g/mol) 410.67
Synonym Octadecanoic acid, 9,10-epoxy-, octyl ester (6CI,7CI,8CI), Oxiraneoctanoic acid, 3-octyl-, octyl ester (9CI), ADK Cizer D 32, Adekacizer D 32, D 32, Drapex 3.2, ED 3, Lankroflex ED 3, Octyl 9,10-epoxystearate, Octyl epoxystearate, Octyl oleate epoxide, 9,10-Epoxystearic Acid, n-Octyl Ester
IUPAC Name octyl 8-(3-octyloxiran-2-yl)octanoate
SMILES CCCCCCCCOC(=O)CCCCCCCC1OC1CCCCCCCC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C26H50O3/c1-3-5-7-9-12-16-20-24-25(29-24)21-17-13-11-14-18-22-26(27)28-23-19-15-10-8-6-4-2/h24-25H,3-23H2,1-2H3
Formula Weight 410.376
Chemical Name or Material Octyl 3-Octyloxiraneoctanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.