missing translation for 'onlineSavingsMsg'
Learn More

Octyl Beta-D-Lactoside, TRC

Product Code. 30413619
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413619 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413619 Supplier Toronto Research Chemicals Supplier No. TRCO28900050MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octyl beta-D-Lactoside
CAS 74513-17-0
Molecular Formula C20 H38 O11
Quantity 50 mg
InChI Formula InChI=1S/C20H38O11/c1-2-3-4-5-6-7-8-28-19-17(27)15(25)18(12(10-22)30-19)31-20-16(26)14(24)13(23)11(9-21)29-20/h11-27H,2-10H2,1H3/t11-,12-,13+,14+,15-,16-,17-,18-,19-,20+/m1/s1
SMILES CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O
IUPAC Name (2S,3R,4S,5R,6R)-2-[(2R,3S,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-octoxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 454.51
Formula Weight 454.2414
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.