missing translation for 'onlineSavingsMsg'
Learn More

Octyl Gallate, LoGiCal

Product Code. 30462044
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30462044 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30462044

Brand: LoGiCal LGCFOR3431.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octylgallate
CAS 1034-01-1
Analyte or Component Names Octylgallate
Shipping Condition Room Temperature
Molecular Formula C15 H22 O5
Certifications/Compliance ISO 9001
Synonym Octyl gallate, Octyl 3,4,5-trihydroxybenzoate, E311
InChI Formula InChI=1S/C15H22O5/c1-2-3-4-5-6-7-8-20-15(19)11-9-12(16)14(18)13(17)10-11/h9-10,16-18H,2-8H2,1H3
Solution Type Neat
SMILES CCCCCCCCOC(=O)c1cc(O)c(O)c(O)c1
IUPAC Name octyl 3,4,5-trihydroxybenzoate
Industry Type Clinical Testing
Molecular Weight (g/mol) 282.33
Quantity 10 mg
Formula Weight 282.1467 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.