missing translation for 'onlineSavingsMsg'
Learn More

Octyl Phenyl Phosphate, TRC

Product Code. 30138507
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138507 1 g 1g
30136680 500 mg 500mg
17982625 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138507 Supplier Toronto Research Chemicals Supplier No. TRCO2938581G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-88-8
Molecular Formula C20 H27 O4 P
Molecular Weight (g/mol) 362.4
Synonym Diphenyl Octyl Phosphate, Octyl diphenyl phosphate
IUPAC Name octyl diphenyl phosphate
SMILES CCCCCCCCOP(=O)(Oc1ccccc1)Oc2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H27O4P/c1-2-3-4-5-6-13-18-22-25(21,23-19-14-9-7-10-15-19)24-20-16-11-8-12-17-20/h7-12,14-17H,2-6,13,18H2,1H3
Formula Weight 362.165
Chemical Name or Material Octyl phenyl phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.